Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139295.3 | NP_647456.1 | 1 - 2 | 111 | 77 | |
GenBank mRNA | AF475283.1 | - | 1 - 2 | 79 | 77 | |
AK033139.1 | - | 1 - 2 | 77 | |||
AK077139.1 | - | 1 - 2 | 62 | 77 | ||
AK077631.1 | - | 1 - 2 | 45 | 77 | ||
AK151803.1 | - | 1 - 2 | 100 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |