Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030133.3 | NP_084409.3 | 11 - 12 | 1558 | 63 | |
GenBank mRNA | BC100359.1 | - | 4 - 5 | 422 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |