Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205234.1 | NP_001192163.1 | 15 - 16 | 3565 | 57 | |
NM_001205235.1 | NP_001192164.1 | 14 - 15 | 3541 | 57 | ||
NM_020253.3 | NP_064649.2 | 15 - 16 | 3565 | 57 | ||
GenBank mRNA | AK129239.1 | - | 15 - 16 | 3538 | 57 | |
AK147295.1 | - | 15 - 16 | 3501 | 57 | ||
AK147665.1 | - | 15 - 16 | 3500 | 57 | ||
BC145496.1 | - | 13 - 14 | 3188 | 9054059 | 57 | |
BC150784.1 | - | 13 - 14 | 3188 | 9087695 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |