Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
NM_001205234.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
NM_001205234.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
NM_001205235.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
NM_001205235.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
NM_020253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
NM_020253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001616.mm.1 |
TC1900000119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
NM_001205234.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
NM_001205234.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
NM_001205235.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
NM_001205235.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
NM_020253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
NM_020253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001617.mm.1 |
TC1900000119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
NM_001205234.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
NM_001205234.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
NM_001205235.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
NM_001205235.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
NM_020253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
NM_020253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001615.mm.1 |
TC1900000119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
NM_001205234.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
NM_001205234.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
NM_001205235.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
NM_001205235.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
NM_020253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
NM_020253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001619.mm.1 |
TC1900000119.mm.1 |
MTA TC |