Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175639.1 | NP_783570.1 | 7 - 8 | 956 | 92 | |
GenBank mRNA | AK012043.1 | - | 7 - 8 | 941 | 92 | |
AK032907.1 | - | 7 - 8 | 930 | 92 | ||
AK129031.1 | - | 7 - 8 | 874 | 92 | ||
AK152321.1 | - | 7 - 8 | 946 | 92 | ||
BC079913.1 | - | 6 - 7 | 657 | 92 | ||
BC117724.2 | - | 8 - 9 | 1161 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |