Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030104.1 | - | 1 - 2 | 2911 | 89 | |
AK122563.1 | - | 1 - 2 | 2924 | 89 | ||
BC059209.1 | - | 1 - 2 | 2930 | 6406239 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |