Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037757.1 | NP_001032846.1 | 5 - 6 | 638 | 66 | |
NM_026833.1 | NP_081109.1 | 5 - 6 | 655 | 66 | ||
GenBank mRNA | AJ304796.1 | - | 5 - 6 | 587 | 66 | |
AK033616.1 | - | 5 - 6 | 632 | 66 | ||
AK149967.1 | - | 5 - 6 | 571 | 66 | ||
AK154400.1 | - | 5 - 6 | 640 | 66 | ||
AK159820.1 | - | 5 - 6 | 583 | 66 | ||
AK171737.1 | - | 5 - 6 | 573 | 66 | ||
AK172673.1 | - | 5 - 6 | 573 | 66 | ||
BC025839.1 | - | 5 - 6 | 568 | 5136195 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |