Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194357.1 | NP_919338.2 | 11 - 12 | 1279 | 61 | |
GenBank mRNA | AK132627.1 | - | 6 - 7 | 631 | 61 | |
BC048084.1 | - | 11 - 12 | 1279 | 6312261 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |