Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194357.1 | NP_919338.2 | 3 - 4 | 413 | 78 | |
GenBank mRNA | BC048084.1 | - | 3 - 4 | 413 | 6312261 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |