Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194357.1 | NP_919338.2 | 10 - 11 | 1108 | 123 | |
GenBank mRNA | AK132627.1 | - | 5 - 6 | 460 | 123 | |
BC048084.1 | - | 10 - 11 | 1108 | 6312261 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |