Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167910.1 | NP_001161382.1 | 11 - 12 | 1886 | 77 | |
NM_019709.4 | NP_062683.3 | 12 - 13 | 2035 | 77 | ||
GenBank mRNA | AF094820.1 | - | 10 - 11 | 1454 | 77 | |
AK029048.1 | - | 11 - 12 | 1863 | 77 | ||
AK122202.1 | - | 11 - 12 | 1852 | 77 | ||
AK147336.1 | - | 10 - 11 | 4455 | 77 | ||
AK147495.1 | - | 11 - 12 | 1860 | 77 | ||
BC054837.1 | - | 10 - 11 | 1654 | 6490394 | 77 | |
BC057198.1 | - | 12 - 13 | 2001 | 5310414 | 77 | |
DQ913734.1 | - | 10 - 11 | 1517 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |