Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054055.2 | NP_473396.1 | 6 - 7 | 948 | 76 | |
GenBank mRNA | AF306491.1 | - | 6 - 7 | 947 | 76 | |
AK085326.1 | - | 6 - 7 | 946 | 76 | ||
AK087518.1 | - | 6 - 7 | 946 | 76 | ||
AK128982.1 | - | 6 - 7 | 945 | 76 | ||
AK138344.1 | - | 6 - 7 | 946 | 76 | ||
AK157881.1 | - | 6 - 7 | 917 | 76 | ||
AK165355.1 | - | 6 - 7 | 947 | 76 | ||
BC026803.1 | - | 6 - 7 | 934 | 4238769 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |