Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081240.3 | NP_001074709.1 | 4 - 5 | 895 | 122 | |
GenBank mRNA | AK045315.1 | - | 4 - 5 | 885 | 122 | |
AK154546.1 | - | 4 - 5 | 839 | 122 | ||
AK205080.1 | - | 2 - 3 | 318 | 122 | ||
AK209748.1 | - | 2 - 3 | 236 | 122 | ||
AK217115.1 | - | 2 - 3 | 261 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |