Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199284.1 | NP_001186213.1 | 1 - 2 | 85 | 78 | |
GenBank mRNA | AB120363.1 | - | 1 - 2 | 81 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |