Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128170.2 | NP_001121642.1 | 12 - 13 | 2264 | 126 | |
NM_001128171.2 | NP_001121643.1 | 12 - 13 | 2389 | 126 | ||
NM_001276279.1 | NP_001263208.1 | 10 - 11 | 1840 | 126 | ||
NM_173369.3 | NP_775545.1 | 11 - 12 | 2255 | 126 | ||
GenBank mRNA | AK122389.1 | - | 12 - 13 | 2370 | 126 | |
AK133479.1 | - | 11 - 12 | 2219 | 126 | ||
AK147204.1 | - | 12 - 13 | 2233 | 126 | ||
AK147426.1 | - | 12 - 13 | 2228 | 126 | ||
AK167424.1 | - | 10 - 11 | 1804 | 126 | ||
BC028885.1 | - | 4 - 5 | 1910 | 126 | ||
BC042438.1 | - | 11 - 12 | 2208 | 3983771 | 126 | |
BC049879.1 | - | 2 - 3 | 117 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |