Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026399.2 | NP_080675.2 | 4 - 5 | 509 | 88 | |
GenBank mRNA | AK005790.1 | - | 4 - 5 | 449 | 88 | |
AK020654.1 | - | 4 - 5 | 88 | |||
AY365434.1 | - | 4 - 5 | 509 | 88 | ||
BC019369.1 | - | 4 - 5 | 432 | 4014928 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |