Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024931.2 | NP_001020102.2 | 12 - 13 | 1384 | 75 | |
NM_001039168.1 | NP_001034257.1 | 12 - 13 | 1381 | 75 | ||
NM_001285438.1 | NP_001272367.1 | 11 - 12 | 1190 | 75 | ||
GenBank mRNA | AK032676.1 | - | 11 - 12 | 1191 | 75 | |
AK049926.1 | - | 12 - 13 | 1363 | 75 | ||
AK086669.1 | - | 12 - 13 | 1367 | 75 | ||
FJ958311.1 | - | 8 - 9 | 798 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |