Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133757.2 | NP_598518.1 | 1 - 2 | 154 | 150 | |
GenBank mRNA | AK046174.1 | - | 1 - 2 | 146 | 150 | |
AK049359.1 | - | 1 - 2 | 155 | 150 | ||
AK053905.1 | - | 1 - 2 | 140 | 150 | ||
AK163967.1 | - | 1 - 2 | 127 | 150 | ||
BC151089.1 | - | 1 - 2 | 254 | 9088002 | 150 | |
BC151185.1 | - | 1 - 2 | 254 | 9088074 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |