Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cse1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023565.3 | NP_076054.1 | 6 - 7 | 649 | 73 | |
GenBank mRNA | AF301152.1 | - | 6 - 7 | 649 | 73 | |
BC145393.1 | - | 6 - 7 | 634 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |