Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139520.1 | NP_001132992.1 | 14 - 15 | 1776 | 70 | |
NM_018851.3 | NP_061339.3 | 14 - 15 | 1776 | 70 | ||
GenBank mRNA | AK054490.1 | - | 14 - 15 | 1675 | 70 | |
AK128954.1 | - | 13 - 14 | 1574 | 70 | ||
AK151335.1 | - | 14 - 15 | 1650 | 70 | ||
AK153390.1 | - | 14 - 15 | 1650 | 70 | ||
AK170688.1 | - | 14 - 15 | 1675 | 70 | ||
AK171565.1 | - | 14 - 15 | 1676 | 70 | ||
BC012721.1 | - | 14 - 15 | 1713 | 4037046 | 70 | |
BC067198.1 | - | 14 - 15 | 1699 | 6826682 | 70 | |
U15635.1 | - | 14 - 15 | 1700 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |