Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019662.2 | NP_062636.2 | 3 - 4 | 705 | 108 | |
GenBank mRNA | AK077271.1 | - | 3 - 4 | 703 | 108 | |
AK087473.1 | - | 3 - 4 | 678 | 108 | ||
AK159550.1 | - | 2 - 3 | 488 | 108 | ||
BC057138.1 | - | 3 - 4 | 684 | 5149047 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |