Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197959.2 | NP_932063.2 | 8 - 9 | 1292 | 84 | |
GenBank mRNA | AK156592.1 | - | 8 - 9 | 1262 | 84 | |
BC057614.1 | - | 8 - 9 | 1260 | 6401536 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |