Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172630.2 | NP_766218.1 | 2 - 3 | 535 | 94 | |
GenBank mRNA | AK041262.1 | - | 2 - 3 | 536 | 94 | |
AK154265.1 | - | 2 - 3 | 389 | 94 | ||
AK158446.1 | - | 2 - 3 | 258 | 94 | ||
AK172000.1 | - | 2 - 3 | 374 | 94 | ||
AK172496.1 | - | 2 - 3 | 290 | 94 | ||
BC045146.1 | - | 2 - 3 | 161 | 6306245 | 94 | |
BC086456.1 | - | 2 - 3 | 208 | 4949311 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |