Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177980.1 | NP_001171451.1 | 4 - 5 | 1190 | 126 | |
NM_001177981.1 | NP_001171452.1 | 4 - 5 | 730 | 126 | ||
NM_001177982.1 | NP_001171453.1 | 4 - 5 | 466 | 126 | ||
NM_019840.2 | NP_062814.2 | 9 - 10 | 1355 | 126 | ||
GenBank mRNA | AF208023.2 | - | 9 - 10 | 1290 | 126 | |
AF326555.1 | - | 9 - 10 | 1095 | 126 | ||
AF326556.1 | - | 9 - 10 | 1095 | 126 | ||
AJ297397.1 | - | 9 - 10 | 1290 | 126 | ||
AK051102.1 | - | 4 - 5 | 731 | 126 | ||
AK150868.1 | - | 4 - 5 | 696 | 126 | ||
AK161341.1 | - | 4 - 5 | 467 | 126 | ||
BC007155.1 | - | 4 - 5 | 537 | 126 | ||
BC023751.1 | - | 4 - 5 | 1166 | 126 | ||
BC071259.1 | - | 4 - 5 | 706 | 5368075 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |