Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017480.2 | NP_059508.2 | 1 - 2 | 100 | 68 | |
GenBank mRNA | AF216748.1 | - | 1 - 2 | 55 | 68 | |
AK030827.1 | - | 1 - 2 | 93 | 68 | ||
BC034852.1 | - | 1 - 2 | 87 | 1364228 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |