Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153556.1 | NP_705784.1 | 9 - 10 | 1947 | 130 | |
GenBank mRNA | AK140689.1 | - | 10 - 11 | 2012 | 130 | |
BC028939.1 | - | 9 - 10 | 1947 | 5364251 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |