Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190984.1 | NP_001177913.1 | 6 - 7 | 771 | 77 | |
NM_001190985.1 | NP_001177914.1 | 5 - 6 | 989 | 77 | ||
NM_021295.3 | NP_067270.1 | 6 - 7 | 805 | 77 | ||
GenBank mRNA | AJ294535.1 | - | 6 - 7 | 798 | 77 | |
AK146120.1 | - | 5 - 6 | 980 | 77 | ||
BC038649.1 | - | 3 - 4 | 310 | 77 | ||
BC058560.1 | - | 6 - 7 | 771 | 6315377 | 77 | |
Y11550.1 | - | 6 - 7 | 770 | 77 | ||
Y16518.1 | - | 5 - 6 | 949 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |