Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039554.3 | NP_001034643.1 | 2 - 3 | 620 | 71 | |
GenBank mRNA | AK031675.1 | - | 2 - 3 | 620 | 71 | |
BC023373.1 | - | 2 - 3 | 607 | 5043159 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |