Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017427.1 | NP_001017427.1 | 5 - 6 | 645 | 105 | |
GenBank mRNA | AK132691.1 | - | 5 - 6 | 625 | 105 | |
BC125539.1 | - | 4 - 5 | 459 | 40130054 | 105 | |
BC137774.1 | - | 4 - 5 | 459 | 8860793 | 105 | |
BC144901.1 | - | 4 - 5 | 459 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |