Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcerg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039474.1 | NP_001034563.1 | 19 - 20 | 2958 | 95 | |
NM_001289526.1 | NP_001276455.1 | 19 - 20 | 2946 | 95 | ||
NR_110344.1 | 19 - 20 | 2958 | 95 | |||
GenBank mRNA | AB023485.1 | - | 19 - 20 | 2922 | 95 | |
AK076993.1 | - | 19 - 20 | 2939 | 95 | ||
AK082750.1 | - | 3 - 4 | 442 | 95 | ||
AK168434.1 | - | 19 - 20 | 2936 | 95 | ||
BC039185.1 | - | 18 - 19 | 2848 | 3982736 | 95 | |
BC040284.1 | - | 18 - 19 | 2858 | 4460736 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |