Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010411.2 | NP_034541.2 | 10 - 11 | 904 | 94 | |
GenBank mRNA | AF074881.1 | - | 10 - 11 | 851 | 94 | |
AF074882.1 | - | 4 - 5 | 278 | 94 | ||
AF098295.1 | - | 10 - 11 | 829 | 94 | ||
AF125536.1 | - | 10 - 11 | 836 | 94 | ||
AK037478.1 | - | 379 | 94 | |||
AK145158.1 | - | 10 - 11 | 870 | 94 | ||
AK206656.1 | - | 3 - 4 | 191 | 94 | ||
BC139300.1 | - | 10 - 11 | 932 | 8862322 | 94 | |
BC139301.1 | - | 10 - 11 | 932 | 8862323 | 94 | |
JN651901.1 | - | 5 - 6 | 303 | 94 | ||
JN651902.1 | - | 4 - 5 | 261 | 94 | ||
JN651903.1 | - | 4 - 5 | 261 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |