Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025590.4 | NP_079866.2 | 12 - 13 | 1357 | 80 | |
GenBank mRNA | AF416923.1 | - | 12 - 13 | 1295 | 80 | |
AK031588.1 | - | 12 - 13 | 1274 | 80 | ||
AK038546.1 | - | 12 - 13 | 1295 | 80 | ||
AK048670.1 | - | 12 - 13 | 1357 | 80 | ||
AK052332.1 | - | 12 - 13 | 1373 | 80 | ||
AK122350.1 | - | 11 - 12 | 1144 | 80 | ||
AK170189.1 | - | 5 - 6 | 380 | 80 | ||
AK171255.1 | - | 5 - 6 | 381 | 80 | ||
BC042492.1 | - | 12 - 13 | 1292 | 4240966 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |