Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029431.1 | NP_083707.1 | 3 - 4 | 499 | 144 | |
GenBank mRNA | AK014834.1 | - | 3 - 4 | 432 | 144 | |
AK038420.1 | - | 3 - 4 | 397 | 144 | ||
AK138397.1 | - | 3 - 4 | 499 | 144 | ||
BC022612.1 | - | 2 - 3 | 347 | 144 | ||
BC134396.1 | - | 3 - 4 | 495 | 40136254 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |