Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005787.1 | NP_001005787.1 | 8 - 9 | 1167 | 72 | |
NM_007704.2 | NP_031730.1 | 8 - 9 | 1167 | 72 | ||
NM_172696.2 | NP_766284.2 | 34 - 35 | 4617 | 72 | ||
GenBank mRNA | AF060539.1 | - | 8 - 9 | 1067 | 72 | |
AK077268.1 | - | 21 - 22 | 2899 | 72 | ||
BC037607.1 | - | 8 - 9 | 839 | 72 | ||
BC057124.1 | - | 8 - 9 | 846 | 72 | ||
BC082787.1 | - | 23 - 24 | 3010 | 72 | ||
BC141404.1 | - | 34 - 35 | 4593 | 9056017 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |