Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172704.3 | NP_766292.2 | 14 - 15 | 1552 | 70 | |
GenBank mRNA | AK053156.1 | - | 14 - 15 | 1534 | 70 | |
AK077544.1 | - | 5 - 6 | 486 | 70 | ||
AK156793.1 | - | 10 - 11 | 4961 | 70 | ||
AK171781.1 | - | 14 - 15 | 1533 | 70 | ||
AK172542.1 | - | 14 - 15 | 1533 | 70 | ||
BC027574.1 | - | 2 - 3 | 202 | 70 | ||
BC075619.1 | - | 11 - 12 | 1645 | 70 | ||
BC079852.1 | - | 14 - 15 | 1522 | 70 | ||
BC085257.1 | - | 14 - 15 | 1526 | 5414963 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |