Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282014.1 | NP_001268943.1 | 5 - 6 | 1352 | 115 | |
NM_001282015.1 | NP_001268944.1 | 5 - 6 | 1346 | 115 | ||
NM_031202.3 | NP_112479.1 | 5 - 6 | 1367 | 115 | ||
GenBank mRNA | AK148332.1 | - | 5 - 6 | 1352 | 115 | |
AK148370.1 | - | 5 - 6 | 1258 | 115 | ||
AK148441.1 | - | 5 - 6 | 1258 | 115 | ||
AK180622.1 | - | 2 - 3 | 270 | 116 | ||
AK190391.1 | - | 2 - 3 | 269 | 115 | ||
AK191129.1 | - | 2 - 3 | 269 | 115 | ||
AK191532.1 | - | 2 - 3 | 269 | 115 | ||
AK193925.1 | - | 2 - 3 | 269 | 115 | ||
AK194181.1 | - | 2 - 3 | 269 | 115 | ||
AK194480.1 | - | 2 - 3 | 269 | 115 | ||
AK194755.1 | - | 2 - 3 | 243 | 115 | ||
AK196212.1 | - | 2 - 3 | 269 | 115 | ||
AK196465.1 | - | 2 - 3 | 269 | 115 | ||
AK197240.1 | - | 2 - 3 | 270 | 116 | ||
AK197957.1 | - | 2 - 3 | 269 | 115 | ||
AK199510.1 | - | 2 - 3 | 269 | 115 | ||
AK199962.1 | - | 2 - 3 | 269 | 115 | ||
AK201297.1 | - | 2 - 3 | 270 | 116 | ||
AK204597.1 | - | 2 - 3 | 269 | 115 | ||
AK205230.1 | - | 2 - 3 | 269 | 115 | ||
AK209301.1 | - | 2 - 3 | 266 | 115 | ||
AK211654.1 | - | 2 - 3 | 269 | 115 | ||
AK219979.1 | - | 2 - 3 | 268 | 115 | ||
BC076598.1 | - | 5 - 6 | 1350 | 30613975 | 115 | |
X03687.1 | - | 5 - 6 | 1252 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |