Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181729.2 | NP_859418.1 | 703 | 95 | ||
GenBank mRNA | BC120907.2 | - | 5 - 6 | 591 | 40087870 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |