Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020623.2 | NP_065648.1 | 1 - 2 | 122 | 80 | |
GenBank mRNA | AK138677.1 | - | 1 - 2 | 120 | 80 | |
BC099456.1 | - | 1 - 2 | 107 | 30845904 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |