Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183180.2 | NP_899003.1 | 5 - 6 | 645 | 72 | |
GenBank mRNA | AK032765.1 | - | 4 - 5 | 556 | 72 | |
BC052169.1 | - | 5 - 6 | 608 | 6335221 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |