Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008914.2 | NP_032940.1 | 10 - 11 | 1317 | 144 | |
GenBank mRNA | AK135203.1 | - | 10 - 11 | 1131 | 144 | |
BC037668.1 | - | 10 - 11 | 1140 | 144 | ||
BC066000.1 | - | 10 - 11 | 1288 | 5699179 | 144 | |
M81483.1 | - | 10 - 11 | 1154 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |