Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207204.2 | NP_997087.2 | 6 - 7 | 890 | 60 | |
GenBank mRNA | AK129253.1 | - | 6 - 7 | 823 | 60 | |
BC067071.1 | - | 6 - 7 | 824 | 30615484 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |