Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019051.mm.1 |
NM_001165892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019051.mm.1 |
NM_001165892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019051.mm.1 |
NM_001282024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019051.mm.1 |
NM_145487.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019051.mm.1 |
NM_145487.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019051.mm.1 |
TC1700001991.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019050.mm.1 |
NM_001165892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019050.mm.1 |
NM_001165892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019050.mm.1 |
NM_001282024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019050.mm.1 |
NM_145487.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019050.mm.1 |
NM_145487.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019050.mm.1 |
TC1700001991.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019053.mm.1 |
NM_001165892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019053.mm.1 |
NM_001165892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019053.mm.1 |
NM_001282024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019053.mm.1 |
NM_145487.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019053.mm.1 |
NM_145487.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019053.mm.1 |
TC1700001991.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_001165892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_001165892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_001282012.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_001282013.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_001282024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_001282025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_145487.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
NM_145487.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019042.mm.1 |
TC1700001991.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019052.mm.1 |
NM_001165892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019052.mm.1 |
NM_001165892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019052.mm.1 |
NM_001282024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019052.mm.1 |
NM_145487.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019052.mm.1 |
NM_145487.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019052.mm.1 |
TC1700001991.mm.1 |
MTA TC |