Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013854.1 | NP_038882.1 | 18 - 19 | 2035 | 69 | |
GenBank mRNA | AF213383.1 | - | 3 - 4 | 297 | 69 | |
AK145549.1 | - | 9 - 10 | 1187 | 69 | ||
AK153057.1 | - | 3 - 4 | 251 | 69 | ||
AK183892.1 | - | 2 - 3 | 136 | 69 | ||
BC040783.1 | - | 7 - 8 | 887 | 69 | ||
BC046965.1 | - | 18 - 19 | 2006 | 6485365 | 69 | |
BC063094.1 | - | 18 - 19 | 2035 | 6516514 | 69 | |
BC068282.1 | - | 2 - 3 | 83 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse ABC Transporters 1700 Inventoried Mouse_ABC_Transporters Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055213 | BC063094.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055213 | BC040783.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055213 | BC046965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055213 | AK145549.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055213 | NM_013854.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | BC063094.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | BC040783.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | BC046965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | AK153057.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | AK145549.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | BC063094.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | BC040783.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | BC046965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | AK153057.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107234 | AK145549.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364196 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965862 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497036 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559051 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94144 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177830 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147040 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | BC063094.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | BC040783.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | BC046965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | AK153057.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | AK145549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | BC063094.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | BC040783.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | BC046965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | AK153057.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107234 | AK145549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055213 | BC063094.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055213 | BC040783.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055213 | BC046965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055213 | AK145549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055213 | NM_013854.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | BC063094.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | BC040783.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | BC046965.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | AK153057.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | AK145549.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | BC063094.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | BC040783.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | BC046965.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | AK153057.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107234 | AK145549.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476008 | AF213383.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476008 | BC063094.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476008 | AK153057.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476008 | AK145549.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476008 | NM_013854.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | BC063094.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | BC040783.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | BC046965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | AK153057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | AK145549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | BC063094.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | BC040783.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | BC046965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | AK153057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107234 | AK145549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P476008 | AF213383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P476008 | BC063094.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P476008 | AK153057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P476008 | AK145549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P476008 | NM_013854.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120843 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13581871 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13582457 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13582458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162023 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450658 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450653 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450665 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450664 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450654 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450667 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450662 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450666 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450668 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450650 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450652 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450660 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450661 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450651 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450655 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450657 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450656 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450663 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450659 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450649 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420158_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452429_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426921_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420157_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452236_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427444_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450648 | - | - |
Mouse Genome 430 2.0 Array | 1420157_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1427444_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420157_s_at | - | - |
Mouse Genome 430 2.0 Array | 1452429_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1452429_s_at | - | - |
Mouse Genome 430 2.0 Array | 1420158_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1426921_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427444_at | - | - |
Mouse Genome 430 2.0 Array | 1420158_s_at | - | - |
Mouse Genome 430 2.0 Array | 1452236_at | - | - |
Mouse Genome 430 2.0 Array | 1426921_at | - | - |
Mouse Genome 430 2.0 Array | 1452236_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224742_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224742_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700018991.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018991.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018991.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700018992.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018992.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018992.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700018995.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018995.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018995.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019004.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019004.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019004.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019006.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019006.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019006.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019008.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019008.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019008.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019001.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019001.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019001.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700018989.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018989.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018989.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700018994.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018994.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018994.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700018999.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018999.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018999.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019007.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019007.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019007.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019009.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019009.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019009.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700018998.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018998.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700018998.mm.1 | TC1700001989.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019005.mm.1 | NM_013854.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019005.mm.1 | NM_013854.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019005.mm.1 | TC1700001989.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452429_PM_s_at | - | - |
Mouse Expression Set 430 | 1452429_s_at | - | - |
Mouse Expression Set 430 | 1427444_PM_at | - | - |
Mouse Expression Set 430 | 1426921_PM_at | - | - |
Mouse Expression Set 430 | 1427444_at | - | - |
Mouse Expression Set 430 | 1452236_at | - | - |
Mouse Expression Set 430 | 1452236_PM_at | - | - |
Mouse Expression Set 430 | 1426921_at | - | - |
Murine Genome U74v2 Set | 97330_PM_at | - | - |
Murine Genome U74v2 Set | 97330_at | - | - |
Murine Genome U74v2 Set | 170555_at | - | - |
Murine Genome U74v2 Set | 170555_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452236_at | - | - |
Mouse Genome 430A 2.0 Array | 1452236_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426921_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427444_at | - | - |
Mouse Genome 430A 2.0 Array | 1452429_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1452429_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1427444_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426921_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427444_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426921_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420158_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427444_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420158_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452429_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426921_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452236_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420157_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452429_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420157_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452236_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 488903 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2760273 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2630707 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3520725 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2760273 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002760273 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002630707 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003520725 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2768926 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693594 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2760415 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2768926 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2693594 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2760415 | - | - |
mouse-6 v1.1 (Illumina) | scl0224742.1_82-S | - | - |
mouse-6 v1.1 (Illumina) | scl49970.18.121_17-S | - | - |
mouse-6 v1.1 (Illumina) | scl00224742.1_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2760273 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3520725 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2630707 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00224742.1_7-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0224742.1_82-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl49970.18.121_17-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760273 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630707 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520725 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2760415 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2693594 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2768926 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2693594 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2760415 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2768926 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0224742.1_82-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl49970.18.121_17-S | - | - |