Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136071.2 | NP_001129543.1 | 1 - 2 | 155 | 63 | |
NM_001271509.1 | NP_001258438.1 | 1 - 2 | 155 | 63 | ||
NM_001271510.1 | NP_001258439.1 | 1 - 2 | 155 | 63 | ||
GenBank mRNA | AK003081.1 | - | 1 - 2 | 104 | 63 | |
BC044736.1 | - | 1 - 2 | 110 | 63 | ||
D49691.1 | - | 1 - 2 | 159 | 63 | ||
M89956.1 | - | 1 - 2 | 97 | 63 | ||
S74179.1 | - | 1 - 2 | 68 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |