Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022315.2 | NP_071710.2 | 10 - 11 | 1219 | 125 | |
GenBank mRNA | AK030958.1 | - | 10 - 11 | 1188 | 125 | |
BC019527.1 | - | 10 - 11 | 1166 | 4191849 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |