Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027452.3 | NP_081728.2 | 1 - 2 | 241 | 81 | |
GenBank mRNA | AK044375.1 | - | 1 - 2 | 118 | 81 | |
AK122476.1 | - | 1 - 2 | 196 | 81 | ||
BC139418.1 | - | 1 - 2 | 241 | 8862440 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |