Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_001165954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_001165954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_001165955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_001165955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_001165956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_001165956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_153421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
NM_153421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013879.mm.1 |
TC0300001792.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_001165954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_001165954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_001165955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_001165955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_001165956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_001165956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_153421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
NM_153421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013880.mm.1 |
TC0300001792.mm.1 |
MTA TC |