Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178038.1 | NP_001171509.1 | 5 - 6 | 925 | 65 | |
GenBank mRNA | BC051402.1 | - | 6 - 7 | 1012 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |