Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028718.2 | NP_082994.1 | 10 - 11 | 1352 | 63 | |
GenBank mRNA | AK014457.1 | - | 7 - 8 | 833 | 63 | |
AY613437.1 | - | 10 - 11 | 1355 | 63 | ||
BC051115.1 | - | 3 - 4 | 226 | 63 | ||
BC117868.2 | - | 10 - 11 | 1326 | 40095476 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |