Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100007922.mm.1 |
NM_001271884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007922.mm.1 |
NM_207031.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007922.mm.1 |
TC0100000933.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007923.mm.1 |
NM_001271884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007923.mm.1 |
NM_207031.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007923.mm.1 |
TC0100000933.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007926.mm.1 |
NM_001271884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007926.mm.1 |
NM_207031.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007926.mm.1 |
TC0100000933.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007924.mm.1 |
NM_001271884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007924.mm.1 |
NM_207031.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007924.mm.1 |
TC0100000933.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007925.mm.1 |
NM_001271884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007925.mm.1 |
NM_207031.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007925.mm.1 |
TC0100000933.mm.1 |
MTA TC |